Would you like to log in again?
SpecCheck Gel Secondary Standard Set, Fluoride, Hach
Supplier: Hach
2712500 2712500
97026-450EA 493.54 USD InStock
97026-450 CA11014-248
SpecCheck Gel Secondary Standard Set, Fluoride, Hach
Standards Environmental Analysis Standards
For Fluoride SPADNS method, 0-2.0 mg/L as F-.
  • Fast and Convenient
  • Stable
Used to confirm consistent instrument response. Set contains four vials filled with gels that simulate the test color at various concentrations.
Ordering information: Set of 4 Vials.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty