Would you like to log in again?
Additional Temperature/RH Datalogger Sensor, Sper Scientific
Supplier: Sper Scientific
800055 800055C
97035-818EA 509.39 USD InStock
97035-818 97035-816
Additional Temperature/RH Datalogger Sensor, Sper Scientific
Data Loggers
Logs up to 16000 readings.
  • Built-in sensor, wall mount and LEDs
  • Both units are supplied with a one year warranty
  • Water resistant
Certified Sensor 97035-818 includes a N.I.S.T. traceable certificate of compliance.
Designed for use with Self-contained dataloggers 97035-812 and 97035-814.
The flexibility of this Datalogger allows you to choose to begin recording immediately, at a predetermined time, or via external activation.
Certifications: CE, RoHS certified.
Caution: LCDs do not work reliably below 20 °C or above 51 °C.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty