Would you like to log in again?
Oxygen Scavenger Reagent Set, Hach
Supplier: Hach
2446600 2446600 2168049
89231-394EA 276.66 USD InStock
89231-394 76441-360 CA24466-00
Oxygen Scavenger Reagent Set, Hach
Environmental Test Kits
For determination of oxygen scavengers by the iron reduction method
Hach method 8140 determines carbohydrazide, diethylhydroxylamine (DEHA), hydroquinone, iso-ascorbic acid (erythorbic acid) and methylethylketoxime.
Ordering information: Set contains DEHA 1 Reagent Powder Pillows and DEHA 2 Solution for 100 tests.
Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accomodate these items.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty