Would you like to log in again?
NitriVer® 2 Nitrite Reagent Powder Pillows, 10 ml, Hach
Supplier: Hach
2107569 2107569
97026-608PK 122.86 USD InStock
CA21075-69 97026-608
NitriVer® 2 Nitrite Reagent Powder Pillows, 10 ml, Hach
Environmental Test Kits
For determination of high range nitrite by the NitriVer® 2 ferrous sulfate method
Hach method 8153. Range: 2 to 250 mg/L.
Ordering information: Pack of 100 Powder Pillows.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty