Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

NitriVer® 3 Nitrite Reagent Powder Pillows, 5 ml, Hach

Supplier: Hach
1407899 1407899
97026-706PK 75.69 USD InStock
CA14078-99 97026-706
NitriVer® 3 Nitrite Reagent Powder Pillows, 5 ml, Hach
Environmental Test Kits
For nitrite determination by the NitriVer® 3 diazotization method

Sample size 5 mL. For use with Hach visual test kits. Range: up to 0.5 mg/L NO2-N.

Ordering information: 100/pk.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty