Would you like to log in again?
Alkalinity Test Kit, Model AL-AP, Hach
Supplier: Hach
2444301 2444301
97026-832EA 143.08 USD InStock
CA11014-752 97026-832
Alkalinity Test Kit, Model AL-AP, Hach
Environmental Test Kits
Caused by the presence of carbonates, bicarbonates, hydroxides, and other dissolved salts, alkalinity is important to food and beverage processors, boiler operators, and aquatic biologists.
  • Simple, economical titration method
  • Rugged kit contains all required reagents, solutions, and glassware
Kit utilizes drop count titration method and contains 100 tests.
*mg/L unless otherwise noted, ppb = μg/L, ppm = mg/L.; gpg = grains per gallon; 1 gpg = 17.1 mg/L or 17.1 ppm. 1EPA-accepted for wastewater. Check with your regulatory agency.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty