Would you like to log in again?
Phenol Reagent Powder Pillows, Hach
Supplier: Hach
87299 87299
89230-890PK 206.84 USD InStock
89230-890 CA872-99
Phenol Reagent Powder Pillows, Hach
Environmental Test Kits
For determination of phenols by the 4-Aminoantipyrine Method
Hach method 8047.
Ordering information: Pack of 100 powder pillows.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty