Would you like to log in again?
BART Test for Fluorescing Pseudomonas, Hach
Supplier: Hach
2432609
97026-612PK 272.01 USD InStock
97026-612
BART Test for Fluorescing Pseudomonas, Hach
Microbiology Test Systems Microorganism Tests
BART biodetectors are excellent diagnostic tools to help identify the presence and aggressivity of various bacteria.
  • Simple yet effective method for monitoring the population size and/or activity of specific groups of bacteria
  • Easy to use, requiring no elaborate or costly equipment and no specialized training
  • Effective and afforable tests are easy to interpret and can be performed at room temperature in virtually any environment
Pseudomonad bacteria are often present in waters that contain oxygen and are rich in organic pollutants (e.g., gasoline, jet fuel, solvents). The presence of pseudomonad bacteria may indicate that aerobic biodegradation is occurring and biofouling may also be happening within the system being tested. Some pseudomonad bacteria that produce the fluorescent pigments (pigments that glow in ultraviolet light) may be a hygiene risk. The faster that clouding and fluorescence happens, the more aggressive are the pseudomonad bacteria. Pseudomonad bacteria can cause a range of problems in water, including slime formations, turbidity, taste and odor, corrosion, biodegradatrion, and hygiene risks. Pseudomonad bacteria produce distinctive odors such as “fishy” or “kerosene-like” odors. In recreational waters (such as swimming pools, hot tubs, restricted natural bathing sites), the presence of aggressive fluorescent pseudomonads can cause various infections.
Ordering information: 9/pk.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty