Would you like to log in again?
Chloride Test Kit, Model CD-DT, Hach
Supplier: Hach
2063500
89230-988PK 787.08 USD InStock
89230-988
Chloride Test Kit, Model CD-DT, Hach
Titrators
The single-parameter Model CD-DT uses the Hach Digital Titrator with Mercuric Nitrate Titrant Cartridges to determine chloride concentrations.
  • Hach Digital Titrator offers higher levels of precision and accuracy than drop count procedures, with reproducibility comparable to a buret titration
  • Rugged titration device is ideal for field use, with no glass burets required
Kit includes Digital Titrator, reagents and labware, manual and carrying case
Contains mercury. Dispose of according to local, state and federal regulations.
Range: 10-8,000 mg/L.
Ordering information: Contains approximately 100 tests.
Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty