Would you like to log in again?
PathoScreen Medium Presence/Absence Powder Pillows, Hach
Supplier: Hach
2610696 2610696
97026-532PK 129.28 USD InStock
97026-532 CA97026-532
PathoScreen Medium Presence/Absence Powder Pillows, Hach
Microbiology Test Systems Microorganism Tests
PathoScreen Medium Presence/Absence Powder Pillows, 100-mL sample.
  • Use to detect waterborne pathogens associated with fecal contamination of water sources
  • Simple presenceabsence method eliminates need for an incubator in tropical and semitropical climates
  • Complete Solutions for Testing Drinking Water
Ordering information: 50/pk.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty