Would you like to log in again?
BART Test for Nitrifying Bacteria, Hach
Supplier: Hach
2619407
97026-484PK 368.96 USD InStock
97026-484
BART Test for Nitrifying Bacteria, Hach
Microbiology Test Systems Microorganism Tests
BART biodetectors are excellent diagnostic tools to help identify the presence and activity of various bacteria.
  • Simple yet effective method for monitoring the population size and/or activity of specific groups of bacteria
  • Easy to use, requiring no elaborate or costly equipment and no specialized training
  • Effective and affordable tests are easy to interpret and can be performed at room temperature in virtually any environment
Nitrifying bacteria recycle organic nitrogenous materials from ammonium (the endpoint for the decomposition of proteins) to nitrates. In water, aggressive nitrifiers can produce high concentrations of nitrates. Nitrates in water can be a potential health risk. Aggressive nitrifying bacteria in waters may indicate the latter stages of aerobic degradation of nitrogen-rich organic matter. This can indicate that the water may have been polluted by nitrogen-rich organics from sources such as compromised septic tanks, sewage systems, industrial and hazardous waste sites and is undergoing an aerobic form of degradation.
Ordering information: 7/pk.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty