Would you like to log in again?
MEL/m-ColiBlue24® Field Filtration Lab, Hach
Supplier: Hach
2586400
89231-414EA 4829.24 USD InStock
89231-414
MEL/m-ColiBlue24® Field Filtration Lab, Hach
Environmental Test Kits
The Coliform and E
  • Consumables and media included
  • EPAapproved field method
m-ColiBlue24® Broth simultaneously detects total coliforms and E. coli in 24 hours. The kit includes all the necessary materials and apparatus for convenient, efficient field operations, and comes in a carrying case for easy storage. Contains: 40 m-ColiBLue24® broth ampules, 150 Push Fit funnels and S-Pak filters (Millipore), 1 Microfil Stand Alone support (Millipore), 1 Syringe/tubing vacuum device, 50 petri dishes (50 mm, sterile) with nutrient pad, 50 sterile, individually wrapped 10 mL pipets (poly), 50 sterile sample bags with dechlorinating agent, alcohol burner and 100 mL of isopropyl alcohol, illuminated magnifier (5x), pipette filler, forceps, ampule breaker and procedure documentation.
coli Field Filtration Kit features manufacturer's m-ColiBlue24® Broth and Millipore's Portable filter assembly, Push Fit funnels and Millipore membranes to simplify membrane filtration testing in the field.
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty