Would you like to log in again?
Differential Pressure Manometer, Model HD700, Extech, Flir
Supplier: Flir
HD700 HD700-NIST
97014-502EA 462.56 USD InStock
97014-502 10171-856
Differential Pressure Manometer, Model HD700, Extech, Flir
Manometers
Low range (0 to 2psi), High Resolution (0.001psi) Gauge/Differential Pressure meter
The HD700 features a large LCD display with backlighting and a heavy duty, double-molded housing. It has functions including Data Hold, Auto power off and a Zero function. Additionally features a built-in USB port.
Certifications: CE marked.
Ordering information: Three year warranty. Windows® compatible software with USB cable,
9V battery, two connection hoses, 100V-240V switching
AC Adaptor, built-in stand, and hard carrying case.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty