Would you like to log in again?
Aluminum Reagent Set, Eriochrome Cyanine R (ECR) Method, 50 ml, Hach
Supplier: Hach
2443400 2443400
89231-086EA 414.12 USD InStock
89231-086 CA24434-00
Aluminum Reagent Set, Eriochrome Cyanine R (ECR) Method, 50 ml, Hach
Environmental Test Kits
For determination of Aluminum by the Eriochrome Cyanine R (ECR) Method
Hach Method 8326. Sample Size: 50 mL Range: 0.002 - 0.220 mg/L. Reagent set contains ECR Reagent Powder Pillows, Hexamethylenetetramine Buffer Reagent Powder Pillows and ECR Masking Reagent Solution for approximately 100 tests.
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty