Would you like to log in again?
MEL/MF Total Coliform Laboratory, Hach
Supplier: Hach
2569700
97026-924EA 7914.55 USD InStock
97026-924
MEL/MF Total Coliform Laboratory, Hach
Environmental Test Kits
The Total Coliform MEL Laboratory is designed for analysts who need portable or small-scale microbiology testing capabilities.
  • Contains: Portable Incubator and rack, Thermometer, Whirlpak bags with thiosulfate (200), Filter funnel/pad assemblies (200), alcohol burner (order alcohol separately), vacuum filtration apparatus, colony counter, magnifier and permanent marker.
  • Choose and order media separately.
From a remote field location to a small scale microbiology lab within your existing lab, this MEL offers all the equipment and accessories to perform microbiological analysis.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty