Would you like to log in again?
BART Test for Blue-Green Algae, Hach
Supplier: Hach
2432727
97026-404PK 1123.73 USD InStock
97026-404
BART Test for Blue-Green Algae, Hach
Microbiology Test Systems Microorganism Tests
BART biodetectors are excellent diagnostic tools to help identify the presence and activity of various bacteria.
  • Simple yet effective method for monitoring the population size and/or activity of specific groups of bacteria
  • Easy to use, requiring no elaborate or costly equipment and no specialized training
  • Effective and affordable tests are easy to interpret and can be performed at room temperature in virtually any environment
Ordering information: 27/pk.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty