Would you like to log in again?
Calcium Standard Solution, 1000 mg/L as Ca (NIST), 500 ml, Hach
Supplier: Hach
2240349 2240349
97021-402EA 85.79 USD InStock
97021-402 CA97021-402
Calcium Standard Solution, 1000 mg/L as Ca (NIST), 500 ml, Hach
Standards Cationic Standards Calcium Standards
For accuracy checks and instrument calibrations in calcium determinations.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty