Would you like to log in again?
Nitrogen-Ammonia Standard Solution, 50 mg/L as NH3-N, 2 ml PourRite™ Ampules, Hach
Supplier: Hach
1479120
97021-796PK 143.67 USD InStock
97021-796
Nitrogen-Ammonia Standard Solution, 50 mg/L as NH3-N, 2 ml PourRite™ Ampules, Hach
Standards Environmental Analysis Standards
For accuracy checks in ammonia nitrogen determinations.
  • Simple Accuracy Checks
  • Minimize waste
  • Reliable results
Ordering information: 20/pk.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty