Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

pH Storage Solution, 3.0 Molar, Hach

Supplier: Hach
2756559
89231-182EA 52.27 USD InStock
89231-182
pH Storage Solution, 3.0 Molar, Hach
Electrolytes Electrode Storage Solutions
Storage solution for pH electrodes.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty