Would you like to log in again?
Gelex Secondary Turbidity Standards Kit, 2100AN, Hach
Supplier: Hach
Gelex
2589200 2589200
97027-026EA 950.3 USD InStock
97027-026 CA25892-00
Gelex Secondary Turbidity Standards Kit, 2100AN, Hach
Standards Turbidity Standards
Gelex Secondary Standards Kit, for 2100AN Laboratory Turbidimeter
Includes: 1 sealed vial each, Stray Light Standard, 0-2 NTU, 0-20 NTU, 0-200 NTU, 200-4000 NTU, 4000-10000 NTU.
Ordering information: 6/pk.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty