Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

CuVer® 1 Copper Reagent Powder Pillows, 5 ml, Hach

Supplier: Hach
1421199 1421199
89230-960PK 60.93 USD InStock
CA11016-918 89230-960
CuVer® 1 Copper Reagent Powder Pillows, 5 ml, Hach
Environmental Test Kits
For determination of copper by the bicinchoninate method

Use with Hach visual test kits. Range: up to 5.0 mg/L Sample Size: 5 mL.

Ordering information: 100/pk.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty