Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-(tert-Butoxycarbonyl)-L-methionine

Supplier: BACHEM AMERICAS INC
Synonyms: Boc-Met-OHBOC-L-MethionineN-Boc-L-methionine

4001022.1000 4001022.0100
A-1935.1000BAEA 1170.52 USD InStock
A-1935.1000BA A-1935.0100BA
N-(tert-Butoxycarbonyl)-L-methionine
N-(tert-Butoxycarbonyl)-L-methionine
Sequence: Boc-Met-OH

Oxidation of Met to the sulfoxide Met(O) can be easily reverted, whereas sulfone (Met(O₂)) formation is practically irreversible.


Formula: C₁₀H₁₉NO₄S
MW: 249.33 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00065586
CAS Number: 2488-15-5
EINECS: 219-639-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty