Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DTXSID30692811

Supplier: BACHEM AMERICAS INC
Synonyms: Fmoc-D-Cys(Trt)-OPfp

4017508.0001
B-2025.0001BAEA 295.67 USD InStock
B-2025.0001BA
DTXSID30692811
DTXSID30692811
Sequence: Fmoc-D-Cys(Trt)-OPfp

Besides "standard" preloaded resins, Bachem offers laterally anchored Fmoc-cysteine, Fmoc-Cys(SASRIN resin)-OH (D-2170) and resin-linked Fmoc-cysteamine, Fmoc-cysteamine-SASRIN resin (D-2165).


Formula: C₄₃H₃₀F₅NO₄S
MW: 751.768 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00153375
CAS Number: 200395-72-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty