Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-Methyl-2-aminopropanoate hydrochloride

Supplier: BACHEM AMERICAS INC
Synonyms: H-Ala-OMe.HClL-Alanine methyl ester hydrochlorideH-Ala-OMe·HCl(S)-2-Aminopropionic Acid Methyl Ester Hydrochloride

4000618.0100 4000618.0025
E-1335.0100BAEA 304.81 USD InStock
E-1335.0100BA E-1335.0025BA
(S)-Methyl-2-aminopropanoate hydrochloride
(S)-Methyl-2-aminopropanoate hydrochloride
Sequence: H-Ala-OMe · HCl

Bachem additionally offers deuterated alanine: Fmoc-[D₄]Ala-OH (B-4130).


Formula: C₄H₉NO₂·HCl
MW: 139.6 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00063663
CAS Number: 2491-20-5
EINECS: 219-652-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty