Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DL-α-Phenylalanine

Supplier: BACHEM AMERICAS INC
Synonyms: DL-Phenylalanine3-Phenyl-DL-alanineH-DL-Phe-OH

4031150.0250
F-1980.0250BAEA 134.17 USD InStock
F-1980.0250BA
DL-α-Phenylalanine
DL-α-Phenylalanine
Sequence: H-DL-Phe-OH

Bachem additionally offers deuterated phenylalanine: Fmoc-[ring-D₅]Phe-OH B-4115.


Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064225
CAS Number: 150-30-1
EINECS: 205-756-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty