Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-Threonine

Supplier: BACHEM AMERICAS INC
Synonyms: (2R,3S)-2-Amino-3-hydroxybutanoic acidH-D-Thr-OH

4031299.0100 4031299.0025
F-2125.0025BAEA 118.93 USD InStock
F-2125.0025BA F-2125.0100BA
D-Threonine
D-Threonine
Sequence: H-D-Thr-OH

Bachem also offers phosphothreonine derivatives.


Formula: C₄H₉NO₃
MW: 119.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064269
CAS Number: 632-20-2
EINECS: 211-171-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty