Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(3-Pyridyl)-L-alanine

Supplier: BACHEM AMERICAS INC
Synonyms: (S)-2-Amino-3-(pyridin-3-yl)propanoic acidH-β-(3-Pyridyl)-Ala-OHH-3-Pal-OH

4014179.0001
F-3195.0001BAEA 396.73 USD InStock
F-3195.0001BA
3-(3-Pyridyl)-L-alanine
3-(3-Pyridyl)-L-alanine
Sequence: H-β-(3-Pyridyl)-Ala-OH
Synonym(s): H-3-Pal-OH#(S)-2-Amino-3-pyridin-3-yl-propionic acid#C³-(3-Pyridyl)-L-alanine

3-Pal derivatives.


Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00079676
CAS Number: 64090-98-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty