Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(R)-2,4-Diamino-4-oxobutanoic acid hydrate

Supplier: BACHEM AMERICAS INC
Synonyms: H-D-Asn-OH·H₂OH-D-Asn-OH(R)-2-Aminosuccinamic acid hydrateD-Asparagine monohydrate

4028945.0025
F-3425.0025BAEA 103.69 USD InStock
F-3425.0025BA
(R)-2,4-Diamino-4-oxobutanoic acid hydrate
(R)-2,4-Diamino-4-oxobutanoic acid hydrate
Sequence: H-D-Asn-OH · H₂O

For isoasparagine derivatives please see the subfamily "Isoasparagine" filed under "Aspartic Acid".

CAS-Number (net): 2058-58-4.


Formula: C₄H₁₀N₂O₄
MW: 150.14 g/mol (132.12 anhy)
Storage Temperature: Ambient
MDL Number: MFCD00149558
CAS Number: 5794-24-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty