Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DL-Aminoglutethimide

Supplier: BACHEM AMERICAS INC
Synonyms: 3-(4-Aminophenyl)-3-ethylpiperidine-2,6-dione

4006364.0005
Q-1020.0005BAEA 609.62 USD InStock
Q-1020.0005BA
DL-Aminoglutethimide
DL-Aminoglutethimide
Sequence: (RS)-3-(4-Amino-phenyl)-3-ethyl-piperidine-2,6-dione
Synonym(s): (RS)-2-(4-Amino-phenyl)-2-ethyl-glutarimide#(RS)-3-(4-Amino-phenyl)-3-ethyl-2,6-dioxo-piperidine

Aminoglutethimide is an inhibitor of steroid aromatase, a key enzyme in adrenocortical steroidogenesis. Aminoglutethimide is applied in the treatment of metastatic breast and prostate cancer, and Cushing's syndrome.


Formula: C₁₃H₁₆N₂O₂
MW: 232.28 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00010122
CAS Number: 125-84-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty