Would you like to log in again?
Iron and pH Color Disc Test Kit, Model IR-23, Hach
Supplier: Hach
256200
89231-282EA 394.68 USD InStock
89231-282
Iron and pH Color Disc Test Kit, Model IR-23, Hach
Environmental Test Kits
Utilizing the simple, accurate color disc method, this Iron & pH Water Quality Test Kit provides accurate, on-site measurement of two important parameters to help maintain water quality within the desired range
Range: 0-10 mg/L/ 4-10 pH unit.
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty