Would you like to log in again?
Nitrogen, Nitrite Color Disc Test Kit, Model NI-6, Hach
Supplier: Hach
224000 224000
89230-934EA 260.65 USD InStock
CA2240-00 89230-934
Nitrogen, Nitrite Color Disc Test Kit, Model NI-6, Hach
Environmental Test Kits
Utilizing the simple color disc method/ferrous sulfate, this kit measures Nitrogen, Nitrite as NO2 and contains 100 tests
  • Simple, lowcost color disc method is the most accurate visual test kit method
  • Kit contains all required reagents and apparatus in a rugged carrying case for analysis in the field
Range (mg/L): 0-100/40-20.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty