Would you like to log in again?
FerroMo™ Iron Reagent 1 Powder Pillows, Hach
Supplier: Hach
2543768 2543768
89231-122PK 68.4 USD InStock
89231-122 CA2543768
FerroMo™ Iron Reagent 1 Powder Pillows, Hach
Environmental Test Kits
For determination of iron in cooling waters containing molybdate-based treatments
Hach Method 8365. Range: 0.01 - 1.80 mg/L Fe.
Ordering information: Pack of 25 powder pillows.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty