Would you like to log in again?
Sodium Thiosulfate Standard Solution, 0.1 N, Hach
Supplier: Hach
32332 32332
97026-380EA 39.45 USD InStock
CA323-32 97026-380
Sodium Thiosulfate Standard Solution, 0.1 N, Hach
Standards Environmental Analysis Standards
For sample dechlorination prior to analysis.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty