Would you like to log in again?
Human Native Transforming growth factor-β (from Phlatelets)
Supplier: Corning
Corning®
354039 356040 356039 356039 356040 354039
47743-608EA 1696.03 USD InStock
CACB-356039A CACB-356040A 47743-608 CACB354039 47743-604 47743-606
Human Native Transforming growth factor-β (from Phlatelets)
Proteins and Peptides
Transforming Growth Factor-beta (TGF-beta) is a multi-functional protein that plays a central role in the regulation of cell growth and differentiation with either stimulatory or inhibitory effects, depending on the context of its action.
Caution: TGF-beta absorbs to glass and plastic surfaces (instructions for dilution are provided with the product).
Tested for ability to inhibit DNA synthesis of Mv 1 Lu mink lung cells (ATCC CCL-64). Tested and found negative for the presence of bacteria, fungi, and mycoplasma.
Source: Human platelets. NOTE: Source material tested for hep B antigen and HIV-1 antibody
Caution: Regulatory status: RUO
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty