Would you like to log in again?
Plate (IMMUNOSET® Pack), Enzo Life Sciences
Supplier: Enzo Life Sciences
80-1930 ADI-80-1930
95052-800EA 136.78 USD InStock
95052-800 CA95052-800
Plate (IMMUNOSET® Pack), Enzo Life Sciences
Microplates Immunology Microplates and Strips
This ImmunoSet plate rack is a convenient set of 5 clear 96 well high-binding polystyrene plates and 5 plate sealers.
Use of these plates will provide optimal performance. The use of different types of plates will produce different results and may affect the sensitivity of the assay due to the type of plastic used.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty