Would you like to log in again?
Leptin (Rat), ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-015A
89141-070EA 763.63 USD InStock
89141-070
Leptin (Rat), ELISA Kit, Enzo Life Sciences
Assays ELISAs
This Leptin (Rat), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours
  • No edge effect: all incubations at room temperature eliminate risk of evaporation at 37 °C
  • Broad dynamic range: easily quantify high and low levels of leptin
  • Convenient method
  • Pre-coated plates and ready-to-use reagents
For the quantitative determination of rat leptin in culture supernatants, plasma, and serum.
Delivery information: Kit contains microtiter plate, standard, antibody, conjugate, assay buffer 28, TMB substrate, stop solution 2, wash buffer concentrate.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty