Would you like to log in again?
Big Endothelin-1 (Human), ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-022
89141-080EA 1005.82 USD InStock
89141-080
Big Endothelin-1 (Human), ELISA Kit, Enzo Life Sciences
Assays ELISAs
The Big Endothelin-1 (Human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results overnight +1 hour.
  • Safe and non-radioactive determination
  • Easy to use
  • Less steps compared to Western blot
  • Reliable and consistent results
For the quantitative determination of human Big ET-1 in culture supernatants and serum. Cited species and sample type include rabbit and lung lavage fluid.
Delivery information: Kit contains microtiter plate, labeled antibody concentrate, antibody diluent, assay buffer, wash buffer concentrate, standard, TMB substrate, stop solution.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty