Would you like to log in again?
GRO/CINC-1 (Rat), ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-074
89141-128EA 1018.32 USD InStock
89141-128
GRO/CINC-1 (Rat), ELISA Kit, Enzo Life Sciences
Assays ELISAs
The GRO/CINC-1 (Rat), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 2 hours
  • Fast and fully quantitative results
  • Sensitive
  • Higher throughput
For the quantitative determination of rat GRO/CINC-1 in culture supernatants, plasma, and serum.
Absorbance is read at 450 nm. Assay up to 39 samples in duplicate per kit.
Delivery information: Supplied with microtiter plate, standard, antibody, conjugate, assay buffer 28, TMB substrate, stop solution 2, wash buffer concentrate.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty