Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IRAK-1 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
905-709-100
89141-360EA 509.17 USD InStock
89141-360
Anti-IRAK-1 Rabbit Polyclonal Antibody
Antibodies
IRAK (IL-1 associated serine/threonine kinase) is a critical regulator of IL-1 induced activation of the NF-κB pathway. IL-1 stimulates the association of IRAK with the heterodimer of IL-IRI and IL-IRAcP via the adapter protein MyD88. In addition to its role in IL-1 signaling, IRAK is a component of the signal transduction pathway through which TNF-R1 activates NF-κB dependent gene expression.

  • For Immunocytochemistry, Immunoprecipitation, Western Blot

Recognizes human, mouse and rat IRAK-1.

Type: Primary
Antigen: IRAK-1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty