Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-15-Deoxy-Delta-12 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
915-044 915-044-100
101106-518EA 464.15 USD InStock
101106-518 CA101106-518
Anti-15-Deoxy-Delta-12 Rabbit Polyclonal Antibody
Antibodies
15-deoxy-Δ12,14-Prostaglandin J2 (15-d-PGJ2) is one of the ultimate dehydration products of PGD2, formed via the intermediate Δ12-PGJ2. 15-d-PGJ2 is physiologically present in body fluids at concentration of 10-12 to 10-9 M, but increases dramatically under stress such as infection and inflammation. 15-d-PGJ2 exerts both pro- and anti-inflammatory effects in a cell-type and concentration-dependent manner, with many of these effects mediated via binding to the γ isoform of the peroxisome proliferators-activated receptor (PPAR-γ). 15-d-PGJ2 appears to induce HO-1 expression by a PPAR-γ-independent mechanism mediated by the generation of reactive oxygen species (ROS).

  • For ELISA, Flow Cytometry

Species independent.

Type: Primary
Antigen: 15-Deoxy-Delta-12
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: pan

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty