Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CGMP Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
915-063-100 915-063
101106-544EA 446.24 USD InStock
101106-544 CA101106-544
Anti-CGMP Rabbit Polyclonal Antibody
Antibodies
Cyclic GMP (cGMP), which has been shown to be present at levels typically 10-100 fold lower than cAMP, is formed by the action of the enzyme guanylate cyclase on GTP. Some hormones, such as Acetylcholine, Insulin, and Oxytocin, as well as certain other chemicals like Serotonin and Histamine cause an increase in cGMP levels. Stimulators of guanylate cyclase such as the vasodilators nitroprusside, nitroglycerin, sodium nitrate, and nitric oxide (NO) also stimulate cGMP levels.

  • For ELISA, Flow Cytometry

Species independent.

Type: Primary
Antigen: CGMP
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty