Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CaN Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
ADI-SPA-610-F SPA-610D ADI-SPA-610-D SPA-610F
95057-956EA 503.54 USD InStock
95057-956 CA95057-954 95057-954 CA95057-956
Anti-CaN Rabbit Polyclonal Antibody
Antibodies
Calcineurin is a Ca2+/Calmodulin-dependent serine/threonine protein phosphatase. The active enzyme is a heterodimer consisting of a catalytic A subunit and a myristylated regulatory B subunit that binds calcium. Calcineurin, which is the target of immunosuppressant drugs such as cyclosporin A and FK506, controls the translocation of Nuclear factors of activated T-cells (NFAT), a family of transcriptional activators that control the expression of cytokine genes essential to immune response, from the cytosol into the nucleus of activated T-cells.

  • For Immunoprecipitation, Western Blot

Recognizes human, mouse, rat, bovine, dog, chicken, Drosophila, hamster, monkey, pig, rabbit and sheep calcineurin A. Detects a band of ~61kDa by Western blot.

Type: Primary
Antigen: CaN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty