Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-KDEL Mouse Monoclonal Antibody [clone: 10C3]

Supplier: Enzo Life Sciences
ADI-SPA-827-J
95046-234EA 2289.61 USD InStock
95046-234
Anti-KDEL Mouse Monoclonal Antibody [clone: 10C3]
Antibodies
The tetrapeptide KDEL, located at the carboxy-terminal sequences of luminal proteins, is a retrieval motif essential for the precise sorting of these proteins along the secretory pathway. KDEL proteins perform essential functions in the endoplasmic reticulum (ER) related to protein folding as well as assembly. The localization of chaperones and other soluble proteins to the ER is achieved by their continuous retrieval from post-ER compartments by the KDEL receptor (Erd2p), which is a membrane protein localized in the Golgi apparatus.

  • For ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot, EM

Recognizes proteins containing the KDEL sequence in mammals, birds, insects, plants, and yeast. Detects three bands of ~94kDa, ~78 kDa and ~55kDa by Western blot..

Type: Primary
Antigen: KDEL
Clonality: Monoclonal
Clone: 10C3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mammals

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty