Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L(+)-Arginine ≥98% (by TLC)

Supplier: Enzo Life Sciences
Synonyms: L-ArginineH-L-Arg-OHArginine(S)-(+)-ArginineH-Arg-OH(S)-2-Amino-5-guanidinopentanoic acid

101-004-G025
89142-488EA 64.4 USD InStock
89142-488
L(+)-Arginine ≥98% (by TLC)
L(+)-Arginine
  • Formulation: White powder.


Formula: C₆H₁₄N₄O₂
MW: 174.2 g/mol
MDL Number: MFCD00002635
CAS Number: 74-79-3
https://us-legacy.vwr.com/stibo/web/std.lang.all/74/16/45347416.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty