Would you like to log in again?
Bovine Native MARCKS Protein (151-175) (from Brain)
Supplier: Enzo Life Sciences
162-004-M001 162-004-MC05
89143-530EA 223.57 USD InStock
89143-530 89143-532
Bovine Native MARCKS Protein (151-175) (from Brain)
Proteins and Peptides
Excellent PKC substrate
Excellent substrate for protein kinase C (PKC) (Km = 20nM for purified rat brain PKC). The peptide consists of residues 151-175 of the myristoylated alanine-rich C-kinase substrate (MARCKS) protein phosphorylation site. Protein kinase C isotypes (including PKCε) phosphorylate serines 8, 12 and 19 of this peptide. It is a very weak substrate for cAMP- and cGMP-dependent protein kinase and CaM kinases I, II and III.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty