Would you like to log in again?
[Ser25]-Protein kinase C (19-31)
Supplier: Enzo Life Sciences
165-017-M005 165-017-M001
89143-696EA 62.32 USD InStock
89143-696 89143-698
[Ser25]-Protein kinase C (19-31)
Enzymes
High affinity PKC substrate.
Pseudosubstrate prototope of the PKC regulatory domain. High affinity substrate for protein kinase C (PKC).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty