Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FURIN Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
210-134-R100
89145-212EA 828.89 USD InStock
89145-212
Anti-FURIN Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Immunocytochemistry, Immunoprecipitation, Western Blot

Recognizes mouse and dog furin convertase and overexpressed human furin convertase. Detects a band of ~100kDa by Western blot. Does not detect endogenous furin from BSC-40, HeLa, J774A-1 BPAEC or CHO cells. Does not cross-react with rat furin convertase.

Type: Primary
Antigen: FURIN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty