Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PPARG2 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

210-192-R100
89145-290EA 798.94 USD InStock
89145-290
Anti-PPARG2 Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Immunocytochemistry, Immunohistochemistry, Western Blot

Recognizes mouse PPARgamma2. Detects a band of ~56kDa by Western blot. Does not cross-react with PPARalpha or PPARdelta.

Type: Primary
Antigen: PPARG2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty