Would you like to log in again?
Z-IETD-pNA
Supplier: Enzo Life Sciences
260-044-M005
89146-676EA 88.09 USD InStock
89146-676
Z-IETD-pNA
Proteins and Peptides
Caspase and granzyme substrate
Fluorogenic substrate for caspase-8 and granzyme B with increased cell permeability. Also substrate for caspase-10.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty