Would you like to log in again?
AC-VDVAD-AFC
Supplier: Enzo Life Sciences
260-112-M010 260-112-M005
89146-866EA 554.78 USD InStock
89146-866 89146-864
AC-VDVAD-AFC
Proteins and Peptides
Caspase-2 substrate
Fluorogenic substrate for caspase-2; similar to Ac-VDVAD-AMC but the AFC fluorophore has a greater Stokes’ shift upon cleavage. VDVAD has been found to be a preferred cleavage site for caspase-2 (Ich-1L). Increases in Ac-VDVAD-AFC cleavage correlated with losses in procaspase-2 (conversion to active caspase-2) in human neuroblastoma lines induced to apoptosis with C2-ceramide and NO.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty